Pierce™ Bovine Serum Albumin, Biotinylated

Catalog number: 29130B

Thermo Scientific™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Biotinylated Proteins include biotin-labeled proteins (BSA), enzymes (HRP, AP) and fluorophores (FITC) for use as controls or signal amplification in IHC via avidin-biotin complex (ABC) techniques.

Biotinylated controls—use biotinylated bovine serum albumin (B-BSA) as a positive control in biotin quantitation methods

Biotin, also known as vitamin H, is a small molecule (MW 244.3) that is present in tiny amounts in all living cells and is critical for a number of biological processes. The valeric acid side chain of the biotin molecule can be derivatized in order to incorporate various reactive groups that are used to attach biotin to other molecules. In the context of immunohistochemistry (IHC), biotin is conjugated to antibodies or to the enzyme reporters used to detect target antigens.

Related Products
Pierce™ Horseradish Peroxidase, Biotinylated
Pierce™ Alkaline Phosphatase, Biotinylated
Pierce™ Biotin-Fluorescein Conjugate
Pierce™ Biotin
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Pierce™
Product Size: Custom

Contents & storage

Store between 2°C and 8°C.

Documents

Manuals & protocols