Search
전체검색
View additional product information for Pierce™ Horseradish Peroxidase, Biotinylated - FAQs (29139)
1 product FAQs found
This biotinylated Horseradish Peroxidase is a native protein with no His-tags.