1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), 96%
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), 96%
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), 96%
Thermo Scientific Chemicals

1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), 96%

CAS: 140681-55-6 | C7H14B2ClF9N2 | 354.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 291490250
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
140.00
Save 20.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
140.00
Save 20.35
Each
Chemical Identifiers
CAS140681-55-6
IUPAC Name4-(chloromethyl)-1-fluoro-1,4-diazabicyclo[2.2.2]octane-1,4-diium; bis(tetrafluoroboranuide)
Molecular FormulaC7H14B2ClF9N2
InChI KeyTXRPHPUGYLSHCX-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Iodometric>=95 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only