Octafluoronaphthalene, 96%
Octafluoronaphthalene, 96%
Octafluoronaphthalene, 96%
Thermo Scientific Chemicals

Octafluoronaphthalene, 96%

CAS: 313-72-4 | C10F8 | 272.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2915300101 gGlass bottle
Catalog number 291530010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS313-72-4
IUPAC Nameoctafluoronaphthalene
Molecular FormulaC10F8
InChI KeyJDCMOHAFGDQQJX-UHFFFAOYSA-N
SMILESFC1=C(F)C2=C(F)C(F)=C(F)C(F)=C2C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
GC>=95 %
Melting point85°C to 90°C
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only