[2,2]-Paracyclophane, 99%
[2,2]-Paracyclophane, 99%
[2,2]-Paracyclophane, 99%
Thermo Scientific Chemicals

[2,2]-Paracyclophane, 99%

CAS: 1633-22-3 | C16H16 | 208.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 291600050
Price (EUR)
69,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
69,00
Each
Chemical Identifiers
CAS1633-22-3
IUPAC Nametricyclo[8.2.2.2⁴,⁷]hexadeca-1(12),4,6,10,13,15-hexaene
Molecular FormulaC16H16
InChI KeyOOLUVSIJOMLOCB-UHFFFAOYSA-N
SMILESC1CC2=CC=C(CCC3=CC=C1C=C3)C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Melting point285°C to 288°C
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only