Sodium triacetoxyborohydride, 97%
Sodium triacetoxyborohydride, 97%
Sodium triacetoxyborohydride, 97%
Thermo Scientific Chemicals

Sodium triacetoxyborohydride, 97%

CAS: 56553-60-7 | C6H10BNaO6 | 211.94 g/mol
数量:
25 g
100 g
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 291821000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS56553-60-7
IUPAC Namesodium bis(acetyloxy)boranuidyl acetate
Molecular FormulaC6H10BNaO6
InChI KeyHHYFEYBWNZJVFQ-UHFFFAOYSA-N
SMILES[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O
さらに表示
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)White
Titration with HClO496.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only