4,4'-Di-tert-butylbiphenyl, 99+%
4,4'-Di-tert-butylbiphenyl, 99+%
4,4'-Di-tert-butylbiphenyl, 99+%
Thermo Scientific Chemicals

4,4'-Di-tert-butylbiphenyl, 99+%

CAS: 1625-91-8 | C20H26 | 266.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 291880050
Price (USD)
55.80
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
55.80
Each
Chemical Identifiers
CAS1625-91-8
IUPAC Name4,4'-di-tert-butyl-1,1'-biphenyl
Molecular FormulaC20H26
InChI KeyCDKCEZNPAYWORX-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(C=C1)C1=CC=C(C=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Melting point126°C to 129°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only