Acetylacetaldehyde dimethyl acetal, 92%
Acetylacetaldehyde dimethyl acetal, 92%
Acetylacetaldehyde dimethyl acetal, 92%
Thermo Scientific Chemicals

Acetylacetaldehyde dimethyl acetal, 92%

CAS: 5436-21-5 | C6H12O3 | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 292330010
Price (USD)
311.00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
311.00
Each
Chemical Identifiers
CAS5436-21-5
IUPAC Name4,4-dimethoxybutan-2-one
Molecular FormulaC6H12O3
InChI KeyPJCCSZUMZMCWSX-UHFFFAOYSA-N
SMILESCOC(CC(C)=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.4130 to 1.4160 (20°C, 589 nm)
Infrared spectrumConforms
GC>=90 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only