Indane, 95%
Indane, 95%
Indane, 95%
Thermo Scientific Chemicals

Indane, 95%

CAS: 496-11-7 | C9H10 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 292480050
Price (USD)
21.65
Special offer
Online exclusive
Ends: 28-Dec-2026
24.80
Save 3.15
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
21.65
Special offer
Online exclusive
Ends: 28-Dec-2026
24.80
Save 3.15
Each
Chemical Identifiers
CAS496-11-7
IUPAC Name2,3-dihydro-1H-indene
Molecular FormulaC9H10
InChI KeyPQNFLJBBNBOBRQ-UHFFFAOYSA-N
SMILESC1CC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5350 to 1.5380 (20°C, 589 nm)
GC>=94 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only