Petroleum ether, extra pure, boiling range 60-95°C
Petroleum ether, extra pure, boiling range 60-95°C
Petroleum ether, extra pure, boiling range 60-95°C
Thermo Scientific Chemicals

Petroleum ether, extra pure, boiling range 60-95°C

CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 292510010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS64742-49-0
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3880 to 1.3920 (20°C, 589 nm)
n-Hexane=<2 %
Color scale=<10 APHA
Boiling point60°C to 95°C
Aromatics=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only