Blood Bank Saline 0.85% - 0.9%
Blood Bank Saline 0.85% - 0.9%
Actual product may vary
Thermo Scientific™

Blood Bank Saline 0.85% - 0.9%

Special processing assures extremely low particle count when you use Fisherbrand™ Blood Bank Saline 0.85% - 0.9%.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantity
293-184Blood Bank Saline, Non-buffered 0.9%1 x 1 gal.
535-435Blood Bank Saline, Non-buffered 0.9%1 x 20 L
312-651Blood Bank Saline, Buffered 0.85%1 x 10 L
309-178Blood Bank Saline, Buffered 0.85%1 x 20 L
062-125Blood Bank Saline, Non-buffered 0.9%1 x 10 L
Catalog number 293-184
Price (TWD)
-
Request A Quote
Description:
Blood Bank Saline, Non-buffered 0.9%
Quantity:
1 x 1 gal.
Special processing assures extremely low particle count when you use Thermo Scientific™ Blood Bank Saline 0.85% - 0.9%, a quality blood bank saline product that is azide- and preservative-free.
  • Consists of a w/v isotonic NaCl solution
  • For in vitro diagnostic and laboratory use only
  • 2.64 gal. (10L) and 5.28 gal. (20L) sizes supplied in Cubitainers™

Non-Buffered Saline

  • W/V: 0.9%
  • pH: 6.0 – 7.5

Buffered Saline

  • W/V: 0.85%
  • pH: 7.2 ± 0.1
  • Phosphate buffered

Note:

Not for infusion, injection or irrigation.

For Laboratory Use
For Laboratory Use
Specifications
DescriptionBlood Bank Saline, Non-buffered 0.9%
Quantity1 x 1 gal.
TypeSaline, Non-Buffered
Unit SizeEach