Hafnium(IV) chloride, 99%, -80 mesh
Hafnium(IV) chloride, 99%, -80 mesh
Hafnium(IV) chloride, 99%, -80 mesh
Thermo Scientific Chemicals

Hafnium(IV) chloride, 99%, -80 mesh

CAS: 13499-05-3 | HfCl4 | 320.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 293251000
Price (USD)
190.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
190.00
Each
Chemical Identifiers
CAS13499-05-3
IUPAC Namehafnium(4+) tetrachloride
Molecular FormulaCl4Hf
InChI KeyPDPJQWYGJJBYLF-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Hf+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration Argentometric98.5 to 101.5 %
Particle size-80 mesh
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only