Thermo Scientific Chemicals

4-Bromo-2-fluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 293920250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromo-2-fluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 293920250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
2105-94-4
IUPAC Name
4-bromo-2-fluorophenol
Molecular Formula
C6H4BrFO
InChI Key
RYVOZMPTISNBDB-UHFFFAOYSA-N
SMILES
OC1=CC=C(Br)C=C1F
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to brown
GC
>=97.5 %
Appearance (Form)
Liquid
Refractive index
1.5660 to 1.5680 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text