3-Bromo-4-fluorobenzaldehyde, 99%
3-Bromo-4-fluorobenzaldehyde, 99%
3-Bromo-4-fluorobenzaldehyde, 99%
Thermo Scientific Chemicals

3-Bromo-4-fluorobenzaldehyde, 99%

CAS: 77771-02-9 | C7H4BrFO | 203.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 293930250
Price (EUR)
88,20
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
88,20
Each
Chemical Identifiers
CAS77771-02-9
IUPAC Name3-bromo-4-fluorobenzaldehyde
Molecular FormulaC7H4BrFO
InChI KeyFAHZIKXYYRGSHF-UHFFFAOYSA-N
SMILESFC1=CC=C(C=O)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point28°C to 33°C
Appearance (Form)Low melting solid
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only