CaptureSelect™ C-tagXL Affinity Matrix
Thermo Scientific™

CaptureSelect™ C-tagXL Affinity Matrix

Thermo Scientific™ CaptureSelect™ C-tagXL Affinity Matrix combines a unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine) with the benefits of a robust and high quality affinity matrix.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
294307201010 mL
29430720055 mL
294307205050 mL
1943072250250 mL
194307201L1 L
194307205L5 L
Número de catálogo 2943072010
Precio (CLP)
-
Cantidad:
10 mL

CaptureSelect C-tagXL Affinity Matrix combines a unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine) with the benefits of a robust and high quality affinity matrix. It purifies C-terminal tagged proteins with high affinity and selectivity, even in the presence of urea and guanidine HCl, from complex mixtures like cell culture harvests and periplasmatic fractions in a one-step process. Mild elution conditions at neutral pH can be applied using magnesium chloride or propylene glycol, which ensures high activity recoveries of pH-sensitive target proteins. C-tagXL Affinity Matrix recognizes the E-P-E-A tag sequence when fused either directly to the C-terminus of a protein or through a linker between the C-terminus and the E-P-E-A tag.

CaptureSelect C-tagXL Affinity Matrix combines a unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine) with the benefits of a robust and high quality affinity matrix. It purifies C-terminal tagged proteins with high affinity and selectivity, even in the presence of urea and guanidine HCl, from complex mixtures like cell culture harvests and periplasmatic fractions in a one-step process. Mild elution conditions at neutral pH can be applied using magnesium chloride or propylene glycol, which ensures high activity recoveries of pH-sensitive target proteins. C-tagXL Affinity Matrix recognizes the E-P-E-A tag sequence when fused either directly to the C-terminus of a protein or through a linker between the C-terminus and the E-P-E-A tag.

Features of C-tagXL Affinity Matrix include:

  • Mild elution, making it suitable for pH-sensitive proteins
  • Binding of the tagged proteins under denaturing conditions (like dissolved inclusion bodies)
  • Excellent scalability
  • Non-animal-derived

CaptureSelect C-tagXL Affinity Matrix purifies C-terminal tagged E-P-E-A proteins directly from complex source materials in a single step with high purity and yield.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect C-tagXL affinity ligand
  • Ligand coupling method: epoxide coupling of the ligand
  • Binding capacity: 400 nmol/mL resin depending on flow rate, column height, and contact time
  • Elution conditions: Acidic: 20 mM citric acid or acetic acid, pH 3–4, or 0.1 M glycine pH 3.0; Neutral: 20 mM Tris, 2.0 M MgCl2 pH 7; 20 mM Tris, 1.0 M NaCl, 50% (v/v) propylene glycol; 20 mM Tris 2 mM S-E-P-E-A.
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Caudal50 to 200 cm/hr. (Up to 2 bar)
Tipo de ligandoC-tagXL affinity ligand
MatrizAgarose-based, epoxide-activated
Cantidad10 mL
Condiciones de envíoTemperatura ambiente
Fase estacionariaAnti-EPEA tag
Capacidad de unión400 nmol/mL
FormularioSuspensión líquida
Tamaño de partículas65 μm
Línea de productosCaptureSelect
TipoAffinity Matrix
Unit SizeEach
Contenido y almacenamiento
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light