CaptureSelect™ tPA Affinity Matrix
Thermo Scientific™

CaptureSelect™ tPA Affinity Matrix

Thermo Scientific™ CaptureSelect™ tPA Affinity Matrix has been specifically designed for the purification of recombinant human tissue plasminogen activator (tPA).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
294343005050 mL
29434300055 mL
294343001010 mL
1943430250250 mL
194343001L1 L
194343005L5 L
Catalog number 2943430050
Price (USD)
-
Quantity:
50 mL

CaptureSelect tPA Affinity Matrix has been specifically designed for the purification of recombinant human tissue plasminogen activator (tPA). This matrix can be used for the purification of recombinant human tPA from cell culture harvests with high purity and yield in a single step. The design of the tPA affinity matrix allows elution of the tPA protein at a neutral pH in the presence of magnesium chloride, but standard low-pH elution buffers like 0.1 M glycine, pH 3.0, can also be used.

Features of this affinity matrix include:

  • Neutral pH elution with 20 mM Tris, 2.0 M MgCl2, 0.2 M arginine pH 7.0
  • Caustic cleaning with 50 mM NaOH
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect tPA affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: 8-10 g/L depending on flow rate, column height, and contact time
  • Elution conditions: Neutral pH: 20 mM Tris, 2.0 M MgCl2, 0.2 M arginine pH 7.0; Low pH: 0.1 M glycine, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity8 to 10 g/L
Flow Rate50 to 200 cm/hr. (Up to 2 bar)
Ligand TypeTPA affinity ligand
MatrixAgarose-based, epoxide-activated
Quantity50 mL
Stationary PhaseAnti-tPA
FormAgarose Bead
Particle Size65 ±10 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).