CaptureSelect™ Bovine IgA Affinity Matrix
Thermo Scientific™

CaptureSelect™ Bovine IgA Affinity Matrix

Thermo Scientific™ CaptureSelect™ Bovine IgA Affinity Matrix has been designed specifically for the purification of bovine IgA from whey.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
29435110055 mL
294351101010 mL
294351105050 mL
Catalog number 2943511005
Price (MXN)
-
Quantity:
5 mL

CaptureSelect Bovine IgA Affinity Matrix has been designed specifically for the purification of bovine IgA from whey.

Features of this affinity matrix include:

  • Binds monomeric, dimeric, and secretory bovine IgA
  • One-step purification from whey
  • Excellent scalability

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect Bovine IgA affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼10 g/L resin
  • Elution conditions: 0.1 M citric acid pH 3.0 or 0.1M glycine pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
Specifications
Binding Capacity∼10 g/L resin
Flow Rate50 to 200 cm/hr. (Up to 2 bar)
Ligand TypeBovine IgA affinity ligand
MatrixAgarose-based, aldehyde-activated
Quantity5 mL
Stationary PhaseAnti Bovine IgA
FormLiquid Suspension
Particle Size65 ±10 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.