CaptureSelect™ IgE Affinity Matrix
Thermo Scientific™

CaptureSelect™ IgE Affinity Matrix

Thermo Scientific™ CaptureSelect™ IgE Affinity Matrix has been designed specifically for the purification of human IgE (immunoglobuline E) from recombinant and plasma sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
294354201010 mL
29435420055 mL
294354205050 mL
1943542250250 mL
194354201L1 L
194354205L5 L
Catalog number 2943542010
Price (USD)
2,095.00
Each
Quantity:
10 mL
Price (USD)
2,095.00
Each

CaptureSelect IgE Affinity Matrix has been designed specifically for the purification of human IgE (immunoglobuline E) from recombinant and plasma sources.

Features of this affinity matrix include:

  • Purification of human IgE, binding different recombinant IgE forms
  • Efficient removal of host cell proteins (HCP) and IgE aggregates
  • Excellent scalability
  • Non–animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect IgE affinity ligand
  • Ligand coupling method: aldehyde coupling
  • Binding capacity: >5 mg/mL resin
  • Elution conditions: 50 mM sodium citrate, 150 mM NaCl, pH 3.5
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity>5 mg/mL Resin
Flow Rate50 to 200 cm/hr. (Up to 2 bar)
Ligand TypeCaptureSelect IgE Affinity Ligand
MatrixAgarose
Max. Operating Pressure2 bar
Quantity10 mL
Shipping ConditionWet Ice
Stationary PhaseIgE
Capacity (Metric)>5 mg/mL Resin
FormResin Slurry
Particle Size65 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C. Protect from light.