CaptureSelect™ TSH Affinity Matrix
Thermo Scientific™

CaptureSelect™ TSH Affinity Matrix

Thermo Scientific™ CaptureSelect™ TSH Affinity Matrix was specifically designed for the purification of thyroid-stimulating hormone (TSH) from recombinant sources.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
294356205050 mL
29435620055 mL
294356201010 mL
1943562250250 mL
194356201L1 L
194356205L5 L
Número de catálogo 2943562050
Precio (MXN)
-
Cantidad:
50 mL

CaptureSelect TSH Affinity Matrix was specifically designed for the purification of thyroid-stimulating hormone (TSH) from recombinant sources.

Some features of CaptureSelect TSH Affinity Matrix include:

  • Highly selective for thyroid-stimulating hormone with no affinity for the other gonadotropins (FSH, LH, and HCG)
  • Enables efficient purification of recombinant TSH with high yields and retained activity
  • Offers excellent scalability
  • Non–animal-derived

The high selectivity and yields obtained using CaptureSelect TSH Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose bead
  • Average particle size: 65 μm
  • Ligand: CaptureSelect TSH affinity ligand
  • Ligand coupling method: epoxide
  • Binding capacity: >5 mg/mL of resin
  • Elution conditions: 0.1 M citrate buffer, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Capacidad de unión>5 mg/mL
Tipo de columnaAgarose Resin
Caudal50 to 200 cm/hr. (Up to 2 bar)
Tipo de ligandoTSH affinity ligand
MatrizAgarose-based
Cantidad50 mL
Condiciones de envíoQualified for Ambient or Wet Ice
Fase estacionariaThyroid Stimulating Hormone (TSH)
ObjetivoThyroid-Stimulating Hormone (TSH)
FormularioAgarose Bead
Tamaño de partículas65 μm
Línea de productosCaptureSelect
TipoAffinity Matrix
Unit SizeEach
Contenido y almacenamiento
Store at 2°C to 8°C.