CaptureSelect™ IgG-Fc (rabbit) Affinity Matrix
CaptureSelect™ IgG-Fc (rabbit) Affinity Matrix
Thermo Scientific™

CaptureSelect™ IgG-Fc (rabbit) Affinity Matrix

Thermo Scientific™ CaptureSelect™ IgG-Fc (Rabbit) Affinity Matrix has been specifically designed for the purification of rabbit IgG from recombinant and plasma sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
294364205050 mL
29436420055 mL
294364201010 mL
Catalog number 2943642050
Price (EUR)
9.210,00
Each
Add to cart
Quantity:
50 mL
Price (EUR)
9.210,00
Each
Add to cart

CaptureSelect IgG-Fc (Rabbit) Affinity Matrix has been specifically designed for the purification of rabbit IgG from recombinant and plasma sources. The ligand on the resin targets the Fc region of rabbit IgG antibodies independent of the light chain subclass.

Features of this affinity matrix include:

  • Highly selective for rabbit IgG
  • No binding to mouse, rat, human, bovine, horse, sheep, and goat IgG
  • Complex growth media with fetal calf or horse serum can be applied
  • Non-animal-derived

The high selectivity and yields obtained using CaptureSelect IgG-Fc (Rabbit) Affinity Matrix enables a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect IgG-Fc (rabbit) affinity ligand
  • Ligand coupling method: epoxide
  • Binding capacity: >15 mg rabbit IgG/mL resin
  • Elution conditions: 1 M glycine, pH 3.0, or 20 mM acetic or citric acid, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity>15 mg rabbit IgG/mL resin
Flow Rate50 to 200 cm/hr. (Up to 2 bar)
Ligand TypeIgG-Fc (rabbit) affinity ligand
MatrixAgarose-based
Quantity50 mL
Shipping ConditionWet Ice
Stationary PhaseAnti-IgG-Fc (rabbit)
FormLiquid Suspension
Particle Size65 ±10 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).