CaptureSelect™ C5 Affinity Matrix
CaptureSelect™ C5 Affinity Matrix
Thermo Scientific™

CaptureSelect™ C5 Affinity Matrix

Thermo Scientific™ CaptureSelect™ C5 Affinity Matrix has been specifically designed for the depletion or purification of human complement component 5 from human plasma.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
294369201010 mL
29436920055 mL
294369205050 mL
카탈로그 번호 2943692010
제품 가격(KRW)
3,622,000
Each
카트에 추가하기
수량:
10 mL
제품 가격(KRW)
3,622,000
Each
카트에 추가하기

CaptureSelect C5 Affinity Matrix has been specifically designed for the depletion or purification of human complement component 5 from human plasma.

Features of this affinity matrix include:

  • Highly selective for human complement component 5
  • Complex growth media with fetal calf serum can be applied
  • Non-animal derived

The high selectivity and yields obtained using CaptureSelect C5 Affinity Matrix enables a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect C5 affinity ligand
  • Ligand coupling method: epoxide
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
사양
유속50 to 200 cm/hr. (Up to 2 bar)
리간드 유형C5 affinity ligand
매트릭스Agarose-based
수량10 mL
배송 조건Wet Ice
정지상C5
형태Agarose Bead
입자 크기65 ±10 μm
제품라인CaptureSelect
유형Affinity Matrix
Unit SizeEach
구성 및 보관
Store in refrigerator (2°C to 8°C).