CaptureSelect™ Factor X Affinity Matrix
Thermo Scientific™

CaptureSelect™ Factor X Affinity Matrix

Thermo Scientific™ CaptureSelect™ Factor X Affinity Matrix was specifically designed for the purification of human Factor X from plasma and recombinant sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
294370201010 mL
29437020055 mL
294370205050 mL
1943702250250 mL
194370201L1 L
194370205L5 L
Catalog number 2943702010
Price (MXN)
-
Quantity:
10 mL

CaptureSelect Factor X Affinity Matrix was specifically designed for the purification of human Factor X from plasma and recombinant sources.

Features of the CaptureSelect Factor X Affinity Matrix include:

  • Highly selective for human Factor X with no affinity for other human plasma proteins
  • Binds both Factor X and Factor Xa in the presence of calcium
  • Enables efficient purification of Factor X with high yields and retained activity
  • Mild elution at neutral pH using EDTA
  • Offers excellent scalability
  • Non-animal-derived

Coagulation Factor X is a vitamin K-dependent, liver-produced serine protease that serves as the first enzyme in the coagulation cascade to form fibrin. It is a two-chain glycoprotein with a molecular weight of approximately 59 kDa. Factor X normally circulates in the plasma as an inactive molecule with activation of Factor X involved in both the intrinsic and extrinsic coagulation pathways.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect Factor X affinity ligand
  • Ligand coupling method: epoxide
  • Binding capacity: >10 mg/mL resin
  • Binding conditions: 50 mM Tris, 150 mM NaCl, 5 mM CaCl2, pH 7
  • Elution conditions: 50 mM Tris, 150 mM NaCl, 5 mM EDTA, pH 7
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity>10 mg/mL
Column TypeAgarose Resin
Flow Rate50 to 200 cm/hr. (Up to 2 bar)
Ligand TypeFactor X affinity ligand
MatrixAgarose-based
Quantity10 mL
Shipping ConditionWet Ice
Stationary PhaseFactor X
TargetProtein
FormAgarose Bead
Particle Size65 ±10 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).