CaptureSelect™ LambdaXP Affinity Matrix
Thermo Scientific™

CaptureSelect™ LambdaXP Affinity Matrix

Thermo Scientific™ CaptureSelect™ LambdaXP Affinity Matrix was specifically designed for the purification of recombinant human immunoglobulins and Fab fragments containing a lambda light chain.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
29437520055 mL
294375201010 mL
294375205050 mL
1943752250250 mL
194375201L1 L
194375205L5 L
194375210L10 L
Número de catálogo 2943752005
Precio (CLP)
-
Cantidad:
5 mL

CaptureSelect LambdaXP Affinity Matrix was specifically designed for the purification of recombinant human immunoglobulins and Fab fragments containing a lambda light chain. The ligand binds to the constant domain of the human lambda light chain and recognizes all isotypes including IgA, IgM, IgD, and IgE.

Features of this affinity matrix include:

  • Efficient elution of Fab fragments and immunoglobulins containing a lambda light chain, no cross binding to the Fc-region or kappa light chains
  • One-step purification from crude material with high purity and yield
  • Excellent scalability
  • Non-animal-derived

CaptureSelect LambdaXP Affinity Matrix can be utilized for the purification of human Ig's and Fab/Fab2 fragments directly from complex source materials and enables a robust and efficient purification process with excellent purity obtained in a single step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect LambdaXP
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >30 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 25 mM sodium acetate, pH 3.5
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
Especificaciones
Caudal50 to 200 cm/hr. (Up to 2 bar)
Para utilizar con (aplicación)Lambda Fab purification
Formulación50% slurry suspension in 20% ethanol
Tipo de ligandoCaptureSelect LambdaXP Affinity Ligand
MatrizAgarose
Presión máx. de funcionamiento2 bar
Cantidad5 mL
Condiciones de envíoWet Ice
ObjetivoLambda light chain
Capacidad de unión>30 g IgG/L Resin
FormularioResin Slurry
Tamaño de partículas65 μm
Línea de productosCaptureSelect
TipoAffinity Matrix
Unit SizeEach
Contenido y almacenamiento
Store at 2°C to 8°C. Protect from light.