CaptureSelect™ AAT-XL Affinity Matrix
Thermo Scientific™

CaptureSelect™ AAT-XL Affinity Matrix

Thermo Scientific™ CaptureSelect™ AAT-XL Affinity Matrix has been specifically designed for the purification of native human alpha-1 antitrypsin (AAT) from complex sources like plasma/serum and recombinant human AAT from cell culture supernatants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
29438320055 mL
294383201010 mL
294383205050 mL
1943832250250 mL
194383201L1 L
194383205L5 L
Catalog number 2943832005
Price (MXN)
-
Quantity:
5 mL

CaptureSelect AAT-XL Affinity Matrix has been specifically designed for the purification of native human alpha-1 antitrypsin (AAT) from complex sources like plasma/serum and recombinant human AAT from cell culture supernatants. This affinity matrix offers mild elution conditions at pH 6. The high selectivity and yield obtained using CaptureSelect AAT-XL Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in a single step.

Features of this affinity matrix include:

  • Highly selective for native and recombinant human AAT
  • High recovery and yield obtained in a single purification step
  • Mild elution at pH 6
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect AAT-XL affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: 15 mg AAT/mL resin
  • Elution conditions: 20 mM Tris, 2.0 M MgCl2, pH 6.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only
Specifications
Flow Rate50 to 200 cm/hr. (Up to 2 bar)
Ligand TypeCaptureSelect Human Alpha-1 Antitrypsin (AAT) Affinity Ligand
MatrixAgarose
Max. Operating Pressure2 bar
Quantity5 mL
Shipping ConditionWet Ice
Binding Capacity15 mg AAT/mL Resin
FormResin Slurry
Particle Size65 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.