Terephthaldehyde monodiethylacetal, 97%, stabilized
Terephthaldehyde monodiethylacetal, 97%, stabilized
Terephthaldehyde monodiethylacetal, 97%, stabilized
Thermo Scientific Chemicals

Terephthaldehyde monodiethylacetal, 97%, stabilized

CAS: 81172-89-6 | C12H16O3 | 208.26 g/mol
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 294630250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS81172-89-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5055 to 1.5085 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only