Product Image
Thermo Scientific™

Fluoro-Max Fluorescent Streptavidin-Coated Particles

Simplify the binding process with a universal binding reagent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
294707010111501 mL
29470701010350100 mL
294707010101505 mL
Catalog number 29470701011150
Price (EUR)
-
Quantity:
1 mL
Simplify the binding process with a universal binding reagent by using Thermo Scientific™ Fluoro-Max Fluorescent Streptavidin-Coated Particles in a variety of applications such as clinical diagnostics, immuno/histological studies and research. With an extremely broad Stokes shift, europium chelate particles help prevent nonspecific fluorescence interference. They are also excellent for quantitative membrane-based rapid assays, heterogeneous assays, luminescent assays, research applications, phagocytosis studies, cell surface markers and pore size determination.

When excited with UV light at its maximal absorbance wavelength of 333 nm, europium chelate particles emit long-lived luminescence at 613 nm, with a lifetime of approximately 0.5 milliseconds, which is about 10,000 to 100,000 times longer than the lifetimes of most fluorophores. This extremely long lifetime allows the application of europium particles to time-resolved fluorescence and can reduce the detection limits of fluorescence assays many orders of magnitude by elimination of background interference from relatively short-lived matrix fluorescence.

  • Made by dyeing Thermo Scientific™ OptiLink™ Carboxylate-Modified particles with europium chelate
  • Specifically designed for membrane or automated fluorometric-based applications
  • Available in standard 0.3 μM diameter
  • Dyed internally to prevent dye leaching and to assure maximum surface immunoreactivity

Fluoro-Max Fluorescent Benefits

  • Bind ligands without dye interference
  • Dye-free surface for coupling
  • High protein binding capacity
  • Readily adsorbs proteins
  • Optimize assays by controlling sensitivity, specificity and stability
  • Optimized acid content

Recommended for:

  • Clinical diagnostics
  • Molecular biology
  • Lateral flow, turbidimetric and slide aggulation assays

 

Specifications
Additive0.05% Sodium Azide
Binding PropertyStreptavidin
Concentration1% solids
ContentsFluorescent red dyed polystyrene microspheres in water
Diameter (Metric) Mean0.3 μm
Diameter (Metric) Nominal0.3 μm
Dye TypeEuropium Chelate
IncludesPackage Insert Sheet containing Certicate of Analysis
Parking AreaBiotin binding of ∼1000pmol/mg
Particle Density1.06g/cm³
Quantity1 mL
StabilityProduct stable for a minimum of 24 months
Surface ModificationStreptavidin
Uniformity<5% CV
ColorWhite
Diameter (Metric)0.3 μm
For Use With (Application)Immunohistological Studies
MaterialPolystyrene or Polystyrene with Coploymer Grafted Surface Coated with Streptavidin
Product TypeFluorescent Streptavidin-Coated Particles
Unit SizeEach
Contents & Storage
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.