Sebacoyl chloride, 92%, technical
Sebacoyl chloride, 92%, technical
Sebacoyl chloride, 92%, technical
Thermo Scientific Chemicals

Sebacoyl chloride, 92%, technical

Sebacoyl chloride, 92%, C10H16Cl2O2, CAS Number-111-19-3, decanedioic acid chloride, sebacoyl chloride, cp, octane-1,8-dicarbonyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, sebacoyl chloride, sebacoylchloride, acmc-1byrk, decanedioyl chloride, 500g, 90% min. (GC) | CAS: 111-19-3 | C10H16Cl2O2 | 239.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
500 g
Packaging:
Glass bottle
Catalog number 294905000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-19-3
IUPAC Namedecanedioyl dichloride
Molecular FormulaC10H16Cl2O2
InChI KeyWMPOZLHMGVKUEJ-UHFFFAOYSA-N
SMILESClC(=O)CCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to golden to brown
AppearanceMay develop some turbidity or precipitate
Refractive index1.4670 to 1.4700 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only