4-Vinylbenzoic acid, 96%
4-Vinylbenzoic acid, 96%
4-Vinylbenzoic acid, 96%
Thermo Scientific Chemicals

4-Vinylbenzoic acid, 96%

CAS: 1075-49-6 | C9H8O2 | 148.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 294910010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1075-49-6
IUPAC Name4-ethenylbenzoic acid
Molecular FormulaC9H8O2
InChI KeyIRQWEODKXLDORP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=95 %
Infrared spectrumConforms
Appearance (Form)Powder
Solubility(10% in Methanol) Clear solution
Appearance (Color)Cream to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only