PADL-PET Transfer Pipet
Molecular Bio Products Inc.

PADL-PET Transfer Pipet

Provides accurate small-volume transfer with flexible design and easy handling.
Have Questions?
Catalog NumberVolume (Metric)
29550 μL
Catalog number 295
Price (USD)
168.57
Online exclusive
421.00
Save 252.43
Case of 4000
Volume (Metric):
50 μL
Price (USD)
168.57
Online exclusive
421.00
Save 252.43
Case of 4000
Provides accurate small-volume transfer with flexible design and easy handling.
  • Flexible design allows controlled dispensing
  • Lightweight and easy to handle
  • Non-sterile for general lab use
  • Supplied in bulk for efficiency
Specifications
Bulb Draw (Metric)0.2 mL
DescriptionPadl-Pet 50μL Drop Size
Diameter (Metric) Outer3.6 mm
GraduatedNo
Packaging TypeBulk 500/Pk 8Pk/Cs 4000/Cs
Quantity500 Pk.
ColorClear
DisposableSingle-use
Length (Metric)12.4 cm
MaterialPolyethylene
SterilityNon-sterile
TypeTransfer Pipet
Volume (Metric)50 μL
Unit SizeCase of 4000