Thermo Scientific™
Plain Microscope Slides
Related applications:
Anatomical Pathology
Experience easy handling with the Thermo Scientific™ Plain Microscope Slides, which feature finely ground edges. These slides are manufactured to the highest quality standards for use in hematology, microbiology, and urinalysis as well as other general laboratory and research applications. All slides are precleaned for immediate use. The surface of each slide is completely plain without a frosted tab.
Experience easy handling with the Thermo Scientific™ Plain Microscope Slides, which feature finely ground edges. These slides are manufactured to the highest quality standards for use in hematology, microbiology, and urinalysis as well as other general laboratory and research applications. All slides are precleaned for immediate use. The surface of each slide is completely plain without a frosted tab.
Back to top
  • Reduced iron content for low self-fluorescence
  • Manufactured from pure, water-white glass
  • Precleaned for immediate use