Thermo Scientific™
Frosted Microscope Slides
 Related applications:
Anatomical Pathology
Easily write on or label a slide with the Thermo Scientific™ Frosted Microscope Slides, featuring a sandblast or etch frosted 3/4″ tab with a smooth frosted finish that can be used for marking specimens or patient identification. The frosted end is easy to write on or can be used for slide labels as well. This feature is available on both sides of the slide with our double-frosted slides. Laboratory applications include Hematology, Microbiology, and Urinalysis.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Smooth frosted tab marking area for effortless writing or printed labels
  • Manufactured from pure, water-white glass
  • Precleaned for immediate use
Documents
Manuals & protocols