Camphene, 75%, remainder mainly alpha-fenchene
Camphene, 75%, remainder mainly alpha-fenchene
Camphene, 75%, remainder mainly alpha-fenchene
Thermo Scientific Chemicals

Camphene, 75%, remainder mainly alpha-fenchene

CAS: 79-92-5 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 295570010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS79-92-5
IUPAC Name2,2-dimethyl-3-methylidenebicyclo[2.2.1]heptane
Molecular FormulaC10H16
InChI KeyCRPUJAZIXJMDBK-UHFFFAOYNA-N
SMILESCC1(C)C2CCC(C2)C1=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
GC>=70 %
Appearance (Form)Low melting crystalline solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only