Azur eosin methylene-blue, pure, solution according to Giemsa
Azur eosin methylene-blue, pure, solution according to Giemsa
Azur eosin methylene-blue, pure, solution according to Giemsa
Thermo Scientific Chemicals

Azur eosin methylene-blue, pure, solution according to Giemsa

CAS: 51811-82-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 295591000
Price (USD)
26.60
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
26.60
Each
Chemical Identifiers
CAS51811-82-6
IUPAC Name7-amino-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
Molecular FormulaC14H14ClN3S
InChI KeyNALREUIWICQLPS-UHFFFAOYSA-N
SMILES[Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infoGiemsa stain powder in MeOH, Glycerol mixture
Appearance (Form)Solution
Appearance (Color)Dark blue to dark purple
UV Lambda max523 to 526 nm (c=0.1, MeOH) Eosine
UV Lambda max646 to 651 nm (c=0.1, MeOH) Methylene blue
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only