Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 295720250
25 L, Metal Drum, Each
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 295720250
25 L, Metal Drum, Each
Quantity
Packaging

Chemical Identifiers

CAS
540-84-1
IUPAC Name
2,2,4-trimethylpentane
Molecular Formula
C8H18
InChI Key
NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES
CC(C)CC(C)(C)C
Infrared spectrum
Conforms
Refractive index
1.3900 to 1.3920 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=99.0 %
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text