Terbium(III) nitrate hexahydrate, 99.99+%, (trace metal basis)
Terbium(III) nitrate hexahydrate, 99.99+%, (trace metal basis)
Terbium(III) nitrate hexahydrate, 99.99+%, (trace metal basis)
Thermo Scientific Chemicals

Terbium(III) nitrate hexahydrate, 99.99+%, (trace metal basis)

CAS: 13451-19-9 | H12N3O15Tb | 453.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 295790020
Price (USD)
66.65
Online Exclusive
74.40
Save 7.75 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
74.40
Save 7.75 (10%)
Each
Chemical Identifiers
CAS13451-19-9
IUPAC Nameterbium(3+) hexahydrate trinitrate
Molecular FormulaH12N3O15Tb
InChI KeyUTCARTSNNKGRTD-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=99.99 % (TREO base)
Appearance (Form)Granular powder
Appearance (Color)White
Total conc. of imp.as REO=<0.01 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only