Cyclopentene, 95+%
Cyclopentene, 95+%
Cyclopentene, 95+%
Thermo Scientific Chemicals

Cyclopentene, 95+%

CAS: 142-29-0 | C5H8 | 68.11 g/mol
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 295901000
Price (JPY)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS142-29-0
IUPAC Namecyclopentene
Molecular FormulaC5H8
InChI KeyLPIQUOYDBNQMRZ-UHFFFAOYSA-N
SMILESC1CC=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4195 to 1.4245 (20°C, 589 nm)
GC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only