Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Thermo Scientific™

Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes

Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.
Catalog NumberVolume (Metric)Material (Membrane)Pore SizeFits Bottle Neck Size (Metric)
296-3345150 mLPES0.45 μm33mm
596-3320150 mLPES0.2 μm33mm
596-4520150 mLPES0.2 μm45mm
296-4545150 mLPES0.45 μm45mm
595-3320500 mLPES0.2 μm33mm
595-4520500 mLPES0.2 μm45mm
295-3345500 mLPES0.45 μm33mm
295-4545500 mLPES0.45 μm45mm
597-33201000 mLPES0.2 μm33mm
597-45201000 mLPES0.2 μm45mm
290-3320150 mLSFCA0.2 μm33mm
290-4520150 mLSFCA0.2 μm45mm
290-3345150 mLSFCA0.45 μm33mm
290-4545150 mLSFCA0.45 μm45mm
291-3320500 mLSFCA0.2 μm33mm
291-4520500 mLSFCA0.2 μm45mm
291-3345500 mLSFCA0.45 μm33mm
291-4545500 mLSFCA0.45 μm45mm
292-33201000 mLSFCA0.2 μm33mm
292-45201000 mLSFCA0.2 μm45mm
Catalog number 296-3345
Price (GBP)
94.90
Case of 12
Volume (Metric):
150 mL
Material (Membrane):
PES
Pore Size:
0.45 μm
Fits Bottle Neck Size (Metric):
33mm
Price (GBP)
94.90
Case of 12

Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.

  • Available in PES and SFCA membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
  • Available with Fit for Purpose Sterile documentation
Specifications
Diameter (Metric)50 mm
Diameter (Metric) Membrane50 mm
Fits Bottle Neck Size (Metric)33mm
No. per Case12
No. per Pack12
TypeBottle Top Filter
Volume (Metric)150 mL
DisposableSingle-use
Filter TypeMembrane Filter
Fit for PurposeSterile
MaterialPolystyrene
Material (Membrane)PES
Pore Size0.45 μm
SterilitySterile
Unit SizeCase of 12

Frequently asked questions (FAQs)

Can I remove the filter membranes from the Nalgene Rapid-Flow Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes?

No, the filter membranes for the bottle top filters are molded in and cannot be removed.