Potassium tris(3,5-dimethyl-1-pyrazolyl)borohydride, 99%
Potassium tris(3,5-dimethyl-1-pyrazolyl)borohydride, 99%
Potassium tris(3,5-dimethyl-1-pyrazolyl)borohydride, 99%
Potassium tris(3,5-dimethyl-1-pyrazolyl)borohydride, 99%
Thermo Scientific Chemicals

Potassium tris(3,5-dimethyl-1-pyrazolyl)borohydride, 99%

CAS: 17567-17-8 | C15H22BKN6 | 336.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2960200505 gGlass bottle
Catalog number 296020050
Price (USD)
190.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
190.00
Each
Chemical Identifiers
CAS17567-17-8
IUPAC Namepotassium tris(3,5-dimethyl-1H-pyrazol-1-yl)boranuide
Molecular FormulaC15H22BKN6
InChI KeyNTWZGFNSHCFHIJ-UHFFFAOYSA-N
SMILES[K+].CC1=NN([BH-](N2N=C(C)C=C2C)N2N=C(C)C=C2C)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or needles
Titration with HClO4>=98.5 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only