2-Nitrophenyl isocyanate, 99%
2-Nitrophenyl isocyanate, 99%
2-Nitrophenyl isocyanate, 99%
Thermo Scientific Chemicals

2-Nitrophenyl isocyanate, 99%

CAS: 3320-86-3 | C7H4N2O3 | 164.12 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 296690050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3320-86-3
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Yellow
Melting point38°C to 41°C
Appearance (Form)Crystalline solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only