Cyclopropyl Carbinol, 99%
Cyclopropyl Carbinol, 99%
Cyclopropyl Carbinol, 99%
Thermo Scientific Chemicals

Cyclopropyl Carbinol, 99%

CAS: 2516-33-8 | C4H8O | 72.11 g/mol
数量:
25 mL
100 mL
250 mL
1 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 296820250
価格(JPY)
-
数量:
25 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2516-33-8
IUPAC Namecyclopropylmethanol
Molecular FormulaC4H8O
InChI KeyGUDMZGLFZNLYEY-UHFFFAOYSA-N
SMILESOCC1CC1
さらに表示
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4290 to 1.4330 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only