(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, > 95% ee, Thermo Scientific Chemicals
(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, > 95% ee, Thermo Scientific Chemicals
(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, > 95% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, > 95% ee, Thermo Scientific Chemicals

CAS: 79200-56-9 | Molecular Formula: C6H7NO | Molecular Weight: 109.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2970400505 gGlass bottle
Catalog number 297040050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS79200-56-9
IUPAC Name(1R,4S)-2-azabicyclo[2.2.1]hept-5-en-3-one
Molecular FormulaC6H7NO
InChI KeyDDUFYKNOXPZZIW-UHNVWZDZSA-N
SMILESO=C1N[C@@H]2C[C@H]1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point92°C to 100°C
HPLC>=97.5 %
Enantiomeric excess>=95 % (HPLC)
Specific optical rotation-550° to -570° (25°C, 589 nm) (c=1, CH2Cl2)
UV(1 % in CH2Cl2) 1 cm cell
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only