(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%
Thermo Scientific Chemicals

(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%

CAS: 49805-30-3 | C6H7NO | 109.13 g/mol
Have Questions?
Catalog NumberQuantity
29706010010 g
Catalog number 297060100
Price (USD)
154.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
154.00
Each
Chemical Identifiers
CAS49805-30-3
IUPAC Name2-azabicyclo[2.2.1]hept-5-en-3-one
Molecular FormulaC6H7NO
InChI KeyDDUFYKNOXPZZIW-UHFFFAOYNA-N
SMILESO=C1NC2CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point55°C to 59°C
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only