Polypeptone™ Peptone
Gibco™

Polypeptone™ Peptone

Polypeptone Peptone is a mixture of peptones made up of equal parts of pancreatic digest of casein and peptic digestRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
29710810 kg
211910454 g
Catalog number 297108
Price (MXN)
-
Quantity:
10 kg
Polypeptone Peptone is a mixture of peptones made up of equal parts of pancreatic digest of casein and peptic digest of animal tissue. Polypeptone Peptone includes the high content of amino acids and small polypeptides characteristic of pancreatic digest of casein and the larger polypeptides characteristic of peptic digest of animal tissue.

Features:
• Animal origin mixture of peptones
• Includes the high content of amino acids

Applications of Polypeptone Peptone include:
• Found to meet nutritional requirements of various bacteria, fungi, and mammalian cells for which a single source of casein or meat peptones has been unsatisfactory

Specifications
Cell TypeBacteria, Fungi, Mammalian
For Use With (Application)Various bacteria, fungi, and mammalian cells
Product LinePolypeptone
Quantity10 kg
SourceAnimal Origin
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.