1,5-Cyclooctadiene, 99%, stabilized
1,5-Cyclooctadiene, 99%, stabilized
1,5-Cyclooctadiene, 99%, stabilized
Thermo Scientific Chemicals

1,5-Cyclooctadiene, 99%, stabilized

CAS: 111-78-4 | C8H12 | 108.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 297120025
Price (TWD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-78-4
IUPAC Name(1Z,5Z)-cycloocta-1,5-diene
Molecular FormulaC8H12
InChI KeyVYXHVRARDIDEHS-QGTKBVGQSA-N
SMILESC1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer-4'-hydroxyphenyl)propionate
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4920 to 1.4950 (20°C, 589 nm)
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only