beta-Ionone, 96%, synthetic
beta-Ionone, 96%, synthetic
beta-Ionone, 96%, synthetic
Thermo Scientific Chemicals

beta-Ionone, 96%, synthetic

CAS: 14901-07-6 | C13H20O | 192.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 297135000
Price (EUR)
189,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
189,00
Each
Chemical Identifiers
CAS14901-07-6
IUPAC Name(3E)-4-(2,6,6-trimethylcyclohex-1-en-1-yl)but-3-en-2-one
Molecular FormulaC13H20O
InChI KeyPSQYTAPXSHCGMF-BQYQJAHWSA-N
SMILESCC(=O)\C=C\C1=C(C)CCCC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=95 %
Specific gravity(25°C) 0.941 to 0.947
Refractive index1.5190 to 1.5215 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only