Brefeldin A, natural
Brefeldin A, natural
Brefeldin A, natural
Thermo Scientific Chemicals

Brefeldin A, natural

CAS: 20350-15-6 | C16H24O4 | 280.36 g/mol
数量:
5 mg
25 mg
パッケージ:
Glass bottle
製品番号(カタログ番号) 297140250
価格(JPY)
-
数量:
25 mg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS20350-15-6
IUPAC Name(1S,6S,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-1H,4H,6H,7H,8H,9H,11aH,12H,13H,14H,14aH-cyclopenta[f]oxacyclotridecan-4-one
Molecular FormulaC16H24O4
InChI KeyKQNZDYYTLMIZCT-PNFJWZTBSA-N
SMILESC[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1
さらに表示
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Melting point201°C to 207°C
HPLC>=98.0 %
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only