2-Amino-5-chlorophenol, 98%
2-Amino-5-chlorophenol, 98%
2-Amino-5-chlorophenol, 98%
Thermo Scientific Chemicals

2-Amino-5-chlorophenol, 98%

CAS: 28443-50-7 | C6H6ClNO | 143.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2972800505 gGlass bottle
Catalog number 297280050
Price (USD)
52.90
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
52.90
Each
Chemical Identifiers
CAS28443-50-7
IUPAC Name2-amino-5-chlorophenol
Molecular FormulaC6H6ClNO
InChI KeyFZCQMIRJCGWWCL-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Infrared spectrumConforms
Melting point152°C to 157°C
GC>=97.5 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only