Hydroxypropyl-beta-cyclodextrin, 97%
Hydroxypropyl-beta-cyclodextrin, 97%
Hydroxypropyl-beta-cyclodextrin, 97%
Acros Organics

Hydroxypropyl-beta-cyclodextrin, 97%

CAS: 128446-35-5 | C44H75O36 | 1180.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 297561000
HSN 29400000
Price (INR)
67,286.00
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
67,286.00
Each
Chemical Identifiers
CAS128446-35-5
Molecular FormulaC44H75O36
SMILESOCC-*.[H][C@@]12O[C@H](CO)[C@@H](O[C@@]3([H])O[C@H](CO)[C@@H](O[C@@]4([H])O[C@H](CO)[C@@H](O[C@@]5([H])O[C@H](CO)[C@@H](O[C@@]6([H])O[C@H](CO)[C@@H](O[C@@]7([H])O[C@H](CO)[C@@H](O[C@@]8([H])O[C@H](CO)[C@@H](O1)[C@H](O)[C@H]8O)[C@H](O)[C@H]7O)[C@H](O)[C@H]6O)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O
Molecular Weight (g/mol)1180.05
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
HPLC>=96.0 % (On dry substance)
NMR2 to 6 (degree of substitution)
NMRanhydroglucopyranose units
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only