2-Phenylquinoline, 99+%
2-Phenylquinoline, 99+%
2-Phenylquinoline, 99+%
Thermo Scientific Chemicals

2-Phenylquinoline, 99+%

CAS: 612-96-4 | C15H11N | 205.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 297990050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS612-96-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Appearance (Form)Crystalline powder or powder or crystals
Infrared spectrumConforms
Melting point81°C to 84°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only