1-Dimethylamino-but-1-en-3-one, 98%
1-Dimethylamino-but-1-en-3-one, 98%
1-Dimethylamino-but-1-en-3-one, 98%
Thermo Scientific Chemicals

1-Dimethylamino-but-1-en-3-one, 98%

CAS: 2802-08-6 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
500 mL
Packaging:
Glass bottle
Catalog number 298020100
Price (USD)
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2802-08-6
IUPAC Name(3E)-4-(dimethylamino)but-3-en-2-one
Molecular FormulaC6H11NO
InChI KeyQPWSKIGAQZAJKS-SNAWJCMRSA-N
SMILESCN(C)C=CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear red-brown to dark brown
Titration with HClO4>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only